Skip to Content
Merck
All Photos(4)

Key Documents

C109207

Sigma-Aldrich

1,5-Cyclooctadiene

contains 50-150 ppm 4-tert-Butylcatechol, 99%

Synonym(s):

1,5-COD, cis-1,5-Cyclooctadiene, COD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12
CAS Number:
Molecular Weight:
108.18
Beilstein:
2036542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

vapor pressure

25.8 mmHg ( 37.7 °C)
6.8 mmHg ( 25 °C)

Quality Level

Assay

99%

form

liquid

autoignition temp.

431 °F

contains

50-150 ppm 4-tert-Butylcatechol as stabilizer
50-150 ppm 4-tert-Butylcatechol

refractive index

n20/D 1.493 (lit.)

bp

149-150 °C (lit.)

mp

−69 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

SMILES string

C1CC=CCCC=C1

InChI

1S/C8H12/c1-2-4-6-8-7-5-3-1/h1-2,7-8H,3-6H2/b2-1-,8-7-

InChI key

VYXHVRARDIDEHS-QGTKBVGQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Bis(Aluminyl)Magnesium: A Source of Nucleophilic or Radical Aluminium-Centred Reactivity.: Investigates 1,5-Cyclooctadiene′s ability to stabilize reactive intermediates in metal complexes, which is crucial for developing new pharmaceutical agents and enhancing ligand efficiency in transition metal catalysis (Griffin et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleonora Cavallari et al.
The journal of physical chemistry. B, 119(31), 10035-10041 (2015-07-15)
Hyperpolarization of (13)C carboxylate signals of metabolically relevant molecules, such as acetate and pyruvate, was recently obtained by means of ParaHydrogen Induced Polarization by Side Arm Hydrogenation (PHIP-SAH). This method relies on functionalization of the carboxylic acid with an unsaturated
Adrian Tlahuext-Aca et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15997-16005 (2014-09-19)
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service