BR782731
BRAND® 12-well Cell Culture Insert, PET-membrane
pore size 0.4 μm, pack of 4 × 9 ea (multipack)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.13
Recommended Products
material
PET membrane
packaging
pack of 4 × 9 ea (multipack)
manufacturer/tradename
BRAND 782731
size
(12 well)
pore size
0.4 μm
Looking for similar products? Visit Product Comparison Guide
General description
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way.
Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types.
With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Overview
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way.
Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types.
With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Overview
- Use in a hanging or standing position
- Works with all common 6-, 12-, or 24-well multi-well plates
- Suitable for 2D and 3D cell cultures
- Surface: cellGrade™ plus
- Sterility Assurance Level (SAL) of 10-6
- PC or PET membrane
- Pore size: from 0.4 μm and 8.0 μm
- Packaged individually or in practical multi-packs
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service