BR781981
BRAND® microplate BRANDplates®, cellGrade
size 384 wells, size 100 μL, flat bottom, white
Synonym(s):
BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade
About This Item
polystyrene
white wells
384 wells
skirt (F-bottom)
Recommended Products
material
flat bottom
polystyrene
white wells
sterility
sterile
feature
lid
skirt (F-bottom)
packaging
pack of 50 ea
pkg of (individually packed)
manufacturer/tradename
BRAND 781981
size
100 μL
384 wells
working volume
100 μL
color
white
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service