BR781378
BRAND® 96-well PCR plate, skirted
white, cut corner A12, low profile
Synonym(s):
PCR, PCR Plates, Plates
About This Item
white
white wells
black coding
low profile
Recommended Products
material
polypropylene
white
white wells
description
D-Nase/R-Nase, DNA-free, suitable for qPCR
sterility
non-sterile
feature
skirt
black coding
low profile
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781378
technique(s)
PCR: suitable
size
96 well
working volume
200 μm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- BRAND® PCR plates are manufactured from high-quality virgin resins for excellent, reproducible results.
- Cleanroom production: High-quality raw materials without lubricants or chemical additives protects the samples from contamination and minimizes the binding of enzymes and nucleic acid to the walls.
- Compatible with most thermal cyclers.
- Ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer.
- The reinforced top plate holds the PCR plate flat even during heating, making it easier to use in automated systems such as pipetting robots.
- Raised rims of wells protect against cross-contamination allowing a reliable seal with the sealing mats tailored to the plates.
- Low-profile 96-well skirted PCR plates are rigid and optimally suited for use with automatic pipetting systems and plate handling.
- BRAND® PCR plates consisting of 96 PCR tubes (0.2 mL), with alphanumeric coding.
- Ultra-thin wall design facilitates rapid and steady heat transfer.
- Smooth interior tube surface minimizes binding of enzymes and nucleic acids.
- Raised rims of wells protect against cross-contamination and provide tight seal particularly in combination with BRAND´s new sealing mats.
- Furthermore these sealing mats can be easily pierced with standard pipette tips.
- The PCR-plates can also be sealed with traditional self-adhesive sealing films.
- Skirted plates are available with bar code on request.
- Suitable for all standard thermal cyclers.
- Compatible with cap strips and sealing films to protect against evaporation and contamination.
- Ideal for PCR and qPCR applications.
- Fully-skirted plates, are ideal for automation due to their rigidity.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service