Skip to Content
Merck
All Photos(1)

Key Documents

BR701330

BRAND® 96-well microplate, U-bottom

round bottom, non-sterile

Synonym(s):

Microplates, Plates, Sample Storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
polypropylene
round bottom
size:
96 well
well maximum volume:
300 μm

material

polypropylene
round bottom

sterility

non-sterile

packaging

pack of 100 ea

manufacturer/tradename

BRAND 701330

size

96 well

well maximum volume

300 μm

binding type

non-treated surface

General description

PP. For volumes up to 300 µl. Compatible with virtually all leading microplate centrifuges. Raised rings around the orifice of each well minimize possible cross-contamination. The plates can be sealed using self-adhesive films, such as DMSO-resistant sealing film (cross-cut) with alphanumeric coding.

Features and Benefits

  • BRAND® 96-well microplate, U-bottom is a deep-well plate ideal for agglutination and other assays requiring stirring and washing of samples.
  • Features a round-shaped well bottom with no edges.
  • Raised rings around the orifice prevent possible cross-contamination.
  • Compatible with a range of microplate centrifuges.
  • Securely sealable with suitable sealing mats or self-adhesive sealing films.
  • Alphanumeric code for accurate identification.
  • Made from high-transparency medical grade polypropylene (PP) that facilitates sample visibility.
  • Chemically resistant to most solvents including DMSO, phenol, and chloroform.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen D Carter et al.
Science advances, 6(14), eaay9572-eaay9572 (2020-04-10)
The endoplasmic reticulum (ER) is a highly dynamic network of membranes. Here, we combine live-cell microscopy with in situ cryo-electron tomography to directly visualize ER dynamics in several secretory cell types including pancreatic β-cells and neurons under near-native conditions. Using
Gefei Chen et al.
Communications biology, 3(1), 32-32 (2020-01-22)
Molecular chaperones play important roles in preventing protein misfolding and its potentially harmful consequences. Deterioration of molecular chaperone systems upon ageing are thought to underlie age-related neurodegenerative diseases, and augmenting their activities could have therapeutic potential. The dementia relevant domain
SeongJun Han et al.
Oncoimmunology, 9(1), 1737368-1737368 (2020-04-22)
Regulatory T cells are integral to the regulation of autoimmune and anti-tumor immune responses. However, several studies have suggested that changes in T cell signaling networks can result in T cells that are resistant to the suppressive effects of regulatory
Rosalie Heilig et al.
Life science alliance, 3(6) (2020-04-30)
Caspase-1 drives a lytic inflammatory cell death named pyroptosis by cleaving the pore-forming cell death executor gasdermin-D (GSDMD). Gsdmd deficiency, however, only delays cell lysis, indicating that caspase-1 controls alternative cell death pathways. Here, we show that in the absence
Kirsi Savijoki et al.
Microorganisms, 8(1) (2020-01-17)
The present study investigated Staphylococcus aureus ATCC25923 surfaceomes (cell surface proteins) during prolonged growth by subjecting planktonic and biofilm cultures (initiated from exponential or stationary cells) to label-free quantitative surfaceomics and phenotypic confirmations. The abundance of adhesion, autolytic, hemolytic, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service