Skip to Content
Merck
All Photos(1)

Key Documents

B59179

Sigma-Aldrich

2-Bromobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCl
CAS Number:
Molecular Weight:
219.46
Beilstein:
508506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.597 (lit.)

bp

245 °C (lit.)

mp

8-10 °C (lit.)

density

1.679 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccccc1Br

InChI

1S/C7H4BrClO/c8-6-4-2-1-3-5(6)7(9)10/h1-4H

InChI key

NZCKTGCKFJDGFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>233.6 °F

Flash Point(C)

> 112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service