Skip to Content
Merck
All Photos(2)

Documents

B56420

Sigma-Aldrich

2-Bromoaniline

98%

Synonym(s):

2-Amino-1-bromobenzene, 2-Aminobromobenzene, 2-Bromobenzenamine, 2-Bromophenylamine, o-Aminobromobenzene, o-Bromoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NH2
CAS Number:
Molecular Weight:
172.02
Beilstein:
742062
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

229 °C (lit.)

mp

24-28 °C (lit.)

density

1.52 g/mL at 25 °C (lit.)

SMILES string

Nc1ccccc1Br

InChI

1S/C6H6BrN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

AOPBDRUWRLBSDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromoaniline is a primary amine that is widely used as a precursor in the synthesis of organobromine compounds.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigating the Effect of Cross-Conjugation Patterns on the Optoelectronic Properties of 7, 7? Isoindigo-Based Materials
Ren S, et al.
Electronics Letters, 12, 3313-3313 (2023)
Jingjing Zhang et al.
Carbohydrate polymers, 234, 115903-115903 (2020-02-20)
In this study, 2-urea-chitosan oligosaccharide derivatives (2-urea-COS derivatives) and 2,6-diurea-chitosan oligosaccharide derivatives (2,6-diurea-COS derivatives) were successfully designed and synthesized via intermediate 2-methoxyformylated chitosan oligosaccharide. All samples were characterized and compared based on FT-IR, 1H NMR spectroscopy, and elemental analysis. The
Jingjing Zhang et al.
Marine drugs, 18(3) (2020-03-20)
Chitosan is an active biopolymer, and the combination of it with other active groups can be a valuable method to improve the potential application of the resultant derivatives in food, cosmetics, packaging materials, and other industries. In this paper, a
Zabiulla et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 95, 419-428 (2017-09-02)
Disrupted redox balance is implicated in multiple pathologies including malignant progression and tumor angiogenesis. In this investigation, we report the design and development of novel and effective ROS detoxifying azo-hydrazone molecules targeting malignant pathologies and neoangiogenesis. A series of azo-derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service