B51801
1,2-Bis(4-pyridyl)ethane
99%
Synonym(s):
Bpa, 4,4′-Ethylenedipyridine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
mp
110-112 °C (lit.)
SMILES string
C(Cc1ccncc1)c2ccncc2
InChI
1S/C12H12N2/c1(11-3-7-13-8-4-11)2-12-5-9-14-10-6-12/h3-10H,1-2H2
InChI key
DQRKTVIJNCVZAX-UHFFFAOYSA-N
Related Categories
General description
1,2-Bis(4-pyridyl)ethane is an N-donor ligand mainly used in the formation of molecular and coordination polymers with metal ions.
Application
1,2-Bis(4-pyridyl)ethane has been used as a flexible bridging ligand in the synthesis of metal-organic frameworks.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Supramolecular architectures and magnetic behavior of coordination polymers from copper (II) carboxylates and 1, 2-bis (4-pyridyl) ethane as a flexible bridging ligand.
Carballo R
Crystal Growth & Design, 7(6), 1069-1077 (2007)
Andjela A Franich et al.
Journal of inorganic biochemistry, 210, 111158-111158 (2020-07-06)
Three new dinuclear palladium(II) complexes with general formula [{Pd(en)Cl}2(μ-L)]2+ (L is pyridine-based bridging ligand 4,4'-bipyridine (4,4'-bipy, 1), 1,2-bis(4-pyridyl)ethane (bpa, 2), 1,2-bis(4-pyridyl)ethylene (bpe, 3) and en is bidentate coordinated ethylenediamine) were synthesized and characterized by elemental microanalyses, NMR (1H and 13C)
Structural analyses and magnetic properties of 3D coordination polymeric networks of nickel (II) maleate and manganese (II) adipate with the flexible 1, 2-bis (4-pyridyl) ethane ligand.
Mukherjee P. S
Inorganic Chemistry, 42(8), 2695-2703 (2003)
Laura Bravo-García et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-16)
Reactions between pyridinic ligands such as 1,2-bis(4-pyridyl)ethane (bpa) and transition metal cations are a very widespread technique to produce extended coordination polymers such as Metal-Organic Frameworks. In combination with a second ligand these systems could present different topologies and behaviors.
Zn-MOFs Containing Flexible ?, ?-Alkane (or Alkene)-Dicarboxylates and 1, 2-Bis (4-pyridyl) ethane Ligands: CO2 Sorption and Photoluminescence.
Hwang I H
Crystal Growth & Design, 13(11), 4815-4823 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service