Skip to Content
Merck
All Photos(1)

Key Documents

ALD00618

Sigma-Aldrich

(+)-PI Reagent

≥95%

Synonym(s):

(2S,3aR,5S,7aR)-7a-methyl-2-((perfluorophenyl)thio)-5-(prop-1-en-2-yl)hexahydrobenzo[d][1,3,2]oxathiaphosphole 2-sulfide, Phosphorus Incorporation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16F5OPS3
CAS Number:
Molecular Weight:
446.46
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

Quality Level

Assay

≥95%

form

crystals

mp

119-124 °C

SMILES string

Fc1c(c(c(c(c1F)F)S[P]2(=S)S[C@H]3[C@](O2)(CC[C@@H](C3)C(=C)C)C)F)F

InChI

1S/C16H16F5OPS3/c1-7(2)8-4-5-16(3)9(6-8)25-23(24,22-16)26-15-13(20)11(18)10(17)12(19)14(15)21/h8-9H,1,4-6H2,2-3H3/t8-,9+,16+,23?/m0/s1

InChI key

CZCFCQOKUBIPHN-DWBMIXFYSA-N

Application

(+)-PI Reagent is an air- and moisture-tolerant, reagent developed in the Baran lab for stereospecific synthesis of chiral phosphines and methylphosphonate nucleotides. The enantiomer, (−)-PI Reagent, is also available.

Learn more about PI and PSI reagents

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmin Xu et al.
Journal of the American Chemical Society, 142(12), 5785-5792 (2020-02-29)
Phosphorus Incorporation (PI, abbreviated Π) reagents for the modular, scalable, and stereospecific synthesis of chiral phosphines and methylphosphonate nucleotides are reported. Synthesized from trans-limonene oxide, this reagent class displays an unexpected reactivity profile and enables access to chemical space distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service