Skip to Content
Merck
All Photos(1)

Key Documents

ALD00506

Sigma-Aldrich

Yu Borylation Ligand

≥95%

Synonym(s):

2,4-dimethoxyquinoline, Montanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
41116155
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

SMILES string

COC1=CC(OC)=NC2=CC=CC=C21

InChI

1S/C11H11NO2/c1-13-10-7-11(14-2)12-9-6-4-3-5-8(9)10/h3-7H,1-2H3

InChI key

XSZBLRGKCBUWRJ-UHFFFAOYSA-N

General description

Yu Borylation Ligand, also referred to as 2,4-dimethoxyquinoline, has been developed by Professor Jin-Quan Yu and coworkers. 2,4-dimethoxyquinoline can be prepared by reacting 2,4-dichloroquinoline with sodium methoxide.

Application

The Yu Borylation Ligand in tandem with bis(pinacolato)diboron (Aldrich 473294) under palladium catalysis can promote direct borylation of C(sp3)-H bonds. Yu and coworkers have displayed this on both alanine-derived amides and cyclopropanes with directing group assistance from the Yu-Wasa Auxiliary (Aldrich 791806).
Yu Borylation Ligand (2,4-Dimethoxyquinoline) may be used as a starting reagent in the synthesis of 2,4 -dimethoxy-3-(3methylbut-2-enyl)quinoline and 4-methoxy-1H-quinolin-2-one. It may also be used in the preparation of isoprenyl derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound-promoted synthesis of novel 2-chloroquinolin-4-pyrimidine carboxylate derivatives as potential antibacterial agents.
Balaji GL, et al.
Research on Chemical Intermediates, 39(4), 1807-1815 (2013)
Regioselective synthesis and biological evaluation of novel bis (2-chloroquinolines).
Rajesh K, et al.
Research on Chemical Intermediates, 39(9), 4259-4267 (2013)
Keith Jones et al.
Organic & biomolecular chemistry, 1(24), 4380-4383 (2003-12-20)
The synthesis of the quinoline alkaloid atanine 6, by selective demethylation of the 2,4-dimethoxyquinoline 11 is presented. An alternative demethylation utilising a thiolate anion leads to the regioisomeric 4-hydroxyquinoline 13.
Quinoline alkaloids. Part XIII. A convenient synthesis of furoquinoline alkaloids of the dictamnine type.
Collins JF, et al.
Journal of the Chemical Society. Perkin Transactions 1, 94-97 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service