Skip to Content
Merck
All Photos(2)

Key Documents

ALD00288

Sigma-Aldrich

Sodium tert-butylsulfinate

Synonym(s):

TBS-Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NaO2S
CAS Number:
Molecular Weight:
144.17
MDL number:
UNSPSC Code:
41106514
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

SMILES string

CC(C)(C)S(O[Na])=O

InChI

1S/C4H10O2S.Na/c1-4(2,3)7(5)6;/h1-3H3,(H,5,6);/q;+1/p-1

InChI key

VOHJOJMSAGPWBW-UHFFFAOYSA-M

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Radical-Based Regioselective C–H Functionalization of Electron-Deficient Heteroarenes: Scope; Tunability; and Predictability

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service