Skip to Content
Merck
All Photos(1)

Key Documents

ALD00108

Sigma-Aldrich

3,6-Bis(methylthio)-1,2,4,5-tetrazine

≥95% (HPLC)

Synonym(s):

3,6-Bis(methylthio)-s-tetrazine, NSC 674352, s-Tetrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4S2
CAS Number:
Molecular Weight:
174.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (HPLC)

form

powder

mp

82-87 °C

storage temp.

−20°C

SMILES string

CSC1=NN=C(SC)N=N1

InChI

1S/C4H6N4S2/c1-9-3-5-7-4(10-2)8-6-3/h1-2H3

InChI key

ROUDTSAXQIBBFZ-UHFFFAOYSA-N

Application

3,6-Bis(methylthio)-1,2,4,5-tetrazine can be used as a versatile building block in the inverse elecron demand hetero Diels-Alder reactions. The Boger Research Group has reported this powerful approach in the synthesis of alkaloids.
It can also used to synthesize:
  • Substituted indoles and indolines derivatives by sequential [4+2] cycloaddition reactions.
  • 3-(Methylthio)-6-(4-morpholinyl)-1,2,4,5-tetrazine by aromatic nucleophilic substitution reaction.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3, 6-Diphenyl-1, 2, 4, 5-tetrazine
Boger DL, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 65(26), 9120-9124 (2001)
Triphenylamine/tetrazine based ?-conjugated systems as molecular donors for organic solar cells
Quinton C, et al.
New. J. Chem., 39(12), 9700-9713 (2015)
Total synthesis of Amaryllidaceae alkaloids utilizing sequential intramolecular heterocyclic azadiene Diels- Alder reactions of an unsymmetrical 1, 2, 4, 5-tetrazine
Boger DL and Wolkenberg SE
The Journal of Organic Chemistry, 65(26), 9120-9124 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service