Skip to Content
Merck
All Photos(1)

Documents

A38606

Sigma-Aldrich

1-Aminoanthracene

technical grade, 90%

Synonym(s):

1-Anthramine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Beilstein:
2209406
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

mp

114-118 °C (lit.)

SMILES string

Nc1cccc2cc3ccccc3cc12

InChI

1S/C14H11N/c15-14-7-3-6-12-8-10-4-1-2-5-11(10)9-13(12)14/h1-9H,15H2

InChI key

YUENFNPLGJCNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Siddlingeshwar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(3), 490-495 (2008-12-06)
The ground state (micro(g)) and the excited state (micro(e)) dipole moments of three substituted anthraquinones, namely 1-aminoanthracene-9,10-dione (AAQ), 1-(methylamino)anthracence-9,10-dione (MAQ) and 1,5-diaminoanthracene-9,10-dione (DAQ) were estimated in various solvents. The dipole moments (micro(g) and micro(e)) were estimated from Lippert, Bakhshiev, Kawski-Chamma-Viallet
N Miled et al.
Biochimica et biophysica acta, 1645(2), 241-246 (2003-02-08)
In human adults, the enzymatic hydrolysis of dietary fat along the digestive tract is sequentially catalyzed by two main enzymes, human gastric lipase (HGL) and human pancreatic lipase (HPL). Both a chemically inhibited form of HPL as well as an
Patricia Nagnan-Le Meillour et al.
Biochimica et biophysica acta, 1794(8), 1142-1150 (2009-05-05)
Structural and molecular dynamics studies have pointed out the role of aromatic residues in the uptake of ligand by porcine odorant-binding protein (pOBP). The shift of Tyr82 from its position during the opening of the binding cavity has been shown
Florence Vincent et al.
European journal of biochemistry, 271(19), 3832-3842 (2004-09-18)
The structure of bovine odorant-binding protein (bOBP) revealed a striking feature of a dimer formed by domain swapping [Tegoni, M., Ramoni, R., Bignetti, E., Spinelli, S. & Cambillau, C. (1996) Nat. Struct. Biol.3, 863-867; Bianchet, M.A., Bains, G., Pelosi, P.
Mariella Parisi et al.
Biochimica et biophysica acta, 1652(2), 115-125 (2003-12-04)
Unfolding and refolding studies on porcine odorant binding protein (pOBP) have been performed at pH 7 in the presence of guanidinium hydrochloride (GdnHCl). Unfolding, monitored by following changes of protein fluorescence and circular dichroism (CD), was found to be a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service