Skip to Content
Merck
All Photos(1)

Key Documents

938904

Sigma-Aldrich

TissueFab® Discrete GelMA

300 bloom, 50% degree of substitution

Synonym(s):

GeIMA, Gelatin Methacrylate, Gelatin methacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C32H48N9O11)n
CAS Number:
MDL number:
UNSPSC Code:
12352202

Quality Level

description

Application: 3D bioprinting, tissue engineering

form

(powder or chunk(s) or fibers)

color

, white to light brown

storage temp.

−20°C

General description

Gelatin methacryloyl, also known as gelatin methacrylate or GelMA, is a methacrylate-functionalized gelatin biomaterial. The gelatin amine groups are chemically modified with methacrylate groups, which when combined with light and a photoinitiator, can be be photopolymerized to form a hydrogel with a three-dimension network that closely mimices the native extracellular matrix (ECM). GelMA is often used as a scaffold for cell culture and tissue regeneration, as it provides a supportive environment for cell attachment, proliferation, and differentation. Its properties, such as mechanical strength, degradation rate, and gelation behavior, can be adjusted by modifying the degree of methacrylation and crosslinking density.

Many GelMAs currently available have a high degree of variation and inconsistency. To address this, the TissueFab(R) Discrete GelMAs have been developed to achieve a specific degree of methacrylation with low batch to batch variability.

Application

  • Tissue engineering - including bone tissue, cartilage tissue, epidermal tissue, and cardiac tissue
  • Regenerative medicine - including injectable tissue constructs and endothelial cell morphogenesis
  • Drug and cell delivery - in the form of microspheres and hydrogels
  • Surface coatings - of medical devices and implants in order to improve biocompatibility and release therapuetic cargos

Features and Benefits

  • Specific degree of methacrylation - a narrow peak of methacrylation allows for better reproducibility, low batch to batch variability, for improved consistency of gel properties and cell behavior.
  • Biocompatibility - GelMA contains RGD sequences, found in collagen and other natural ECM proteins, which promote cell adhesion, proliferation, differentiation, and maturation of a variety of cell types.
  • Biodegradability - GelMA contains matrix metalloproteinase (MMP) degradable sites that can be recognized and enzymatically degraded by cells. This allows the encapsulated cells to degrade and remodel the GelMA matrix, and repopulate with their own cells and tissues, a major goal for tissue engineering and regenerative medicine.
  • Tunability - The TissueFab(R) Discrete GelMAs are available in a variety of degrees of substitution which ultimately impact the hydrogel stiffness and mechanical properties and can be used to recapitulate and accomodate different native tissues.
  • Bioprintability - The TissueFab(R) Discrete GelMAs are widely used in the field of 3D bioprinting due to their unique gelation properties and the ability to print via extrusion and other methods to create intricate cell-encapsulated 3D structures with high cell viability.

Legal Information

TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Directed endothelial cell morphogenesis in micropatterned gelatin methacrylate hydrogels.
Nikkhah M, Eshak N, Zorlutuna P, et al.
Biomaterials, 33(35), 9009-9018 (2012)
Mineralized gelatin methacrylate-based matrices induce osteogenic differentiation of human induced pluripotent stem cells.
Kang H, Shih YR, Hwang Y, et al.
Acta Biomaterialia, 10(12), 4961-4170 (2014)
Tissue engineering of retina through high resolution 3-dimensional inkjet bioprinting.
Masaeli, et al.
Biofabrication, 12, 25006-25006 (2020)
Photocrosslinkable gelatin hydrogel for epidermal tissue engineering.
Zhao X, et al.
Advanced Helathcare Materials (2015)
Facile one-step micropatterning using photodegradable methacrylated gelatin hydrogels for improved cardiomyocyte organization and alignment.
Tsang K, et al.
Advances in Functional Materials, 25(6), 977-986 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service