928208
Gold nanoparticles
90 nm diameter, azide functionalized, 1 ML, OD 50, in water
Synonym(s):
AuNP, GdNP, Gold NP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
description
Alkyne surface density: 1/nm2
concentration
in water
OD
50
core diam.
90 nm , Coefficient of Variance < 12%
diameter
90 nm
λmax
564 nm
PDI
<0.200
functional group
azide
storage temp.
2-8°C
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Azide functionalized gold nanoparticles are suitable for covalent conjugation of any molecule with a terminal alkyne through click chemistry. These gold nanoparticles allow for rapid generation of gold conjugates for subsequent use in downstream assays.
Azide functionalized gold nanoparticles are more than 95% spherical and have uniform size distribution (CV <12%).
Azide functionalized gold nanoparticles are more than 95% spherical and have uniform size distribution (CV <12%).
Application
Ideal for development of gold conjugates for use in applications such as blotting, lateral flow assays, microscopy, and transmission electron microscopy (TEM).
Storage and Stability
When stored for a long period of time gold nanoparticles may sediment at the bottom of the vial, which is especially prominent for larger particle sizes. Prior to use, re-suspend the sedimented particles by thorough mixing until a homogenous solution is obtained.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioconjugate chemistry, 17(6), 1373-1375 (2006-11-16)
A simple and versatile method for the preparation of functional enzyme-gold nanoparticle conjugates using "click" chemistry has been developed. In a copper-catalyzed 1,2,3-triazole cycloaddition, an acetylene-functionalized Thermomyces lanuginosus lipase has been attached to azide-functionalized water-soluble gold nanoparticles under retention of
Beilstein journal of organic chemistry, 14, 11-24 (2018-01-31)
Glycoscience, despite its myriad of challenges, promises to unravel the causes of, potential new detection methods for, and novel therapeutic strategies against, many disease states. In the last two decades, glyco-gold nanoparticles have emerged as one of several potential new
Chemical communications (Cambridge, England), (2), 169-171 (2007-12-20)
We present a new type of azide-functionalized gold nanoparticle and their coupling to an alkyne-modified DNA duplex using the copper(I)-catalyzed Huisgen cycloaddition ('click chemistry'), resulting in a chain-like assembly of nanoparticles on the DNA template.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service