Skip to Content
Merck
All Photos(3)

Key Documents

926302

Sigma-Aldrich

Snowman Mixer Chip

Fluidic 1108, COC

Synonym(s):

Microfluidic chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

description

Microfludic chip x1

Application

The Snowman Mixer Fluidic 1108 COC microfluidic chip applies passive mixing principles and its unique shape to achieve fast mixing of two solutions.

Features and Benefits

  • Eight identical micromixing units per chip
  • Each unit features one outlet two inlets and one outlet
  • Instantaneous mixing is achieved by adding small volumes (up to 5 μl) of Solution A and larger volumes of solution B which are immediately transferred and mixed in the large reaction chamber
  • Ideal for signal read-out of bioreactions (fluorescence, luminescence, etc.) due to fast mixing times
  • This chip is not meant to be used in flow-through mode

Components

Chip Properties:
  • Mini Luer Interface
  • Material: Cyclic olefin copolymer (COC)
  • Reaction chamber volume: 50 μl

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service