Skip to Content
Merck
All Photos(1)

Key Documents

924016

Sigma-Aldrich

Silver bromide

≥99.99% trace metals basis

Synonym(s):

Argentous bromide, Silver monobromide, Silver(I) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgBr
CAS Number:
Molecular Weight:
187.77
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
≥99.99% trace metals basis
form:
powder, crystals or chunks

Quality Level

Assay

≥99.99% trace metals basis

form

powder, crystals or chunks

mp

432 °C (lit.)

density

6.473 g/mL at 25 °C (lit.)

SMILES string

Br[Ag]

InChI

1S/Ag.BrH/h;1H/q+1;/p-1

InChI key

ADZWSOLPGZMUMY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver bromie is a photosensitive material used as photocatalyst. A study reports hydrogen generation using AgBr as a photocatatlyst in CH3OH/H2O solution. AgBr doped aerogels have shown enhanced photocatalystic and antibacterial activities.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Double Z-scheme system of silver bromide@ bismuth tungstate/tungsten trioxide ternary heterojunction with enhanced visible-light photocatalytic activity
Zhang M, et al.
Journal of Colloid and Interface Science, 509, 18-24 (2018)
Silver bromide as a photocatalyst for hydrogen generation from CH3OH/H2O solution
Kakuta N, et al.
The Journal of Physical Chemistry B, 103(29), 5917-5919 (1999)
Fabrication of a three-dimensional porous Z-scheme silver/silver bromide/graphitic carbon nitride@ nitrogen-doped graphene aerogel with enhanced visible-light photocatalytic and antibacterial activities
Chen Y, et al.
Journal of Colloid and Interface Science, 536, 389-398 (2019)
Recent advances in silver bromide-based Z-scheme photocatalytic systems for environmental and energy applications: A review
Sharma S, et al.
Journal of Environmental Chemical Engineering, 9, 105157-105157 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service