Skip to Content
Merck
All Photos(1)

Key Documents

920134

Sigma-Aldrich

Graphene, monolayer film

6 in diameter, CVD on silicon wafer, large grain, avg. no. of layers, 1

Synonym(s):

Roll-to-Roll graphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12141908
NACRES:
NA.23

product name

R2R Monolayer large grain CVD graphene on silicon wafer, 6 in diameter, avg. no. of layers, 1

Quality Level

description

Growth method: roll-to-roll CVD
Wafer: SiO2 (300nm) Si
Number of layer: Monolayer
Raman intensity 2D/G: ≥1.5

feature

avg. no. of layers 1

sheet resistance

280 Ω/sq ±10%

size

110 μm × 110 μm ± 10%, grain size

surface coverage

surface coverage >98%

transmittance

>97%

semiconductor properties

(mobility>3000 cm2/V·s) (Hall effect measurements)

Looking for similar products? Visit Product Comparison Guide

General description

Roll-to-roll, high-quality, monolayer CVD graphene with large grain size (~110μm^2) on 6 inch silicon wafer.

Application

Our Roll-to-Roll CVD graphene products are true monolayer high quality graphene, fabricated inside a Cleanroom, heavily monitored and QC to assure high reproducibility.
The roll-to-roll process allows continuous, large scale graphene production.

This large grain size graphene product on silicon wafer is ready to use, helps you minimize process time, and increase success rate. This product with low sheet resistance would enable unmatched reproducibility and allow high performance for CVD graphene based FET, CVD graphene based sensors, and heterostructure based micro/nano electronics.

Application examples:
  • Ultrafast Transistor
  • Optical devices
  • Bio/Gas Sensor
  • Transparent Electrode
  • Flexible Display
  • Smart Coating
  • Thermal management

Caution

Be cautious not to drop
Keep away from contamination, heat, dust and flame etc.

Storage and Stability

Avoid direct sun light, avoid high temperature, avoid high humidity, and avoid dust.

Legal Information

Product of LG Electronics, R&D use only

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Ling Tang et al.
ACS nano, 11(12), 12817-12823 (2017-11-29)
Two-dimensional (2D) materials are drawing growing attention for next-generation electronics and optoelectronics owing to its atomic thickness and unique physical properties. One of the challenges posed by 2D materials is the large source/drain (S/D) series resistance due to their thinness
Integrating graphene into semiconductor fabrication lines.
Daniel Neumaier et al.
Nature materials, 18(6), 525-529 (2019-05-23)
Bing Deng et al.
Advanced materials (Deerfield Beach, Fla.), 31(9), e1800996-e1800996 (2018-10-03)
Chemical vapor deposition (CVD) is considered to be an efficient method for fabricating large-area and high-quality graphene films due to its excellent controllability and scalability. Great efforts have been made to control the growth of graphene to achieve large domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service