Skip to Content
Merck
All Photos(1)

Key Documents

91660

Sigma-Aldrich

Tridodecylamine

≥97.0% (GC)

Synonym(s):

Trilaurylamine, Hydrogen ionophore I

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N
CAS Number:
Molecular Weight:
521.99
Beilstein:
1801669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

≥97.0% (GC)
≥97.0% (NT)

form

liquid

refractive index

n20/D 1.4578 (lit.)
n20/D 1.458

bp

220-228 °C/0.03 mmHg (lit.)

density

0.823 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C36H75N/c1-4-7-10-13-16-19-22-25-28-31-34-37(35-32-29-26-23-20-17-14-11-8-5-2)36-33-30-27-24-21-18-15-12-9-6-3/h4-36H2,1-3H3

InChI key

SWZDQOUHBYYPJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tridodecylamine(TDA) can be used in the following applications:
  • Extraction of picolinic acid, citric acid, silver, mercury, selenium, zinc, cobalt, iron and succinic acid from aqueous solutions by using various organic solvents as diluents.
  • Recovery of Remazol Red 3BS (R3BS) dye from wastewater via emulsion liquid membrane (ELM) process by using TDA as the carrier agent.
  • Synthesis of gemini surfactant-based ionic liquids by reacting with dibromo alkanes.

Caution

may form precipitate on storage

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

374.0 °F

Flash Point(C)

190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction equilibria of picolinic acid from aqueous solution by tridodecylamine (TDA)
Tuyun AF and Uslu H
Desalination, 268(1-3), 134-140 (2011)
Extraction of certain elements from aqueous methanol, ethanol and acetone by tridodecylamine and tributyl phosphate
Alian A, et al.
Talanta, 15(2), 249-255 (1968)
Recovery of synthetic dye from simulated wastewater using emulsion liquid membrane process containing tri-dodecyl amine as a mobile carrier
Othman N, et al.
Journal of Hazardous Materials, 198(5), 103-112 (2011)
Simple one step synthesis of gemini cationic surfactant-based ionic liquids: Physicochemical, surface properties and biological activity
Tawfik SM
Journal of Molecular Liquids, 209(5), 320-326 (2015)
Comparative equilibrium studies for citric acid by amberlite LA-2 or Tridodecylamine (TDA)
Bayazit SS, et al.
Journal of Chemical and Engineering Data, 54(7), 1991-1996 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service