Skip to Content
Merck
All Photos(1)

Documents

913626

Sigma-Aldrich

Silica nanoparticles

pore size non-porous, 700 nm particle size, 5 % (w/w) in ethanol, dispersion

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

5 % (w/w) in ethanol

particle size

700 nm

pore size

non-porous

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging whereas micron sized particles are used in liquid chromatography. We offer highly monodisperse nonporous silica in ethanol, with sizes varying from 150 nm to 900 nm. The high monodispersity of our particles makes them suitable for aforementioned applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Tang et al.
Nano today, 8(3), 290-312 (2013-09-03)
Nanomedicine, the use of nanotechnology for biomedical applications, has potential to change the landscape of the diagnosis and therapy of many diseases. In the past several decades, the advancement in nanotechnology and material science has resulted in a large number
Martha Kim et al.
Scientific reports, 7(1), 8238-8238 (2017-08-16)
Nonporous silica nanoparticles (SiNPs) have potential as promising carriers for ophthalmic drugs. However, the in vivo safety of ocular topical SiNPs remains unclear. This study investigated the in vivo safety of oral and ocular topical applications of 100 nm-sized SiNPs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service