Skip to Content
Merck
All Photos(1)

Documents

908576

Sigma-Aldrich

WinterGreen photocage

≥95%

Synonym(s):

(1,3,5,5,7,9-Hexamethyl-dipyrrolo[1,3,2]diazaborinin-10-yl)methanol, 4,4′-Dimethyl-8-hydroxymethyl-1,3,5,7-tetramethyl-4-bora-3a,4a-diaza-s-indacene, BODIPY photocage, Imidazole-protected BODIPY alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23BN2O
CAS Number:
Molecular Weight:
270.18

Assay

≥95%

form

powder or crystals

storage temp.

2-8°C

Application

Green-light absorbing photocage. Easily functionalized to ester with DCC/EDC couplings for photorelease of carboxylic acids.

Legal Information

US Patent Application #16/140,218

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratik P Goswami et al.
Journal of the American Chemical Society, 137(11), 3783-3786 (2015-03-10)
Photoremovable protecting groups derived from meso-substituted BODIPY dyes release acetic acid with green wavelengths >500 nm. Photorelease is demonstrated in cultured S2 cells. The photocaging structures were identified by our previously proposed strategy of computationally searching for carbocations with low-energy
Tomáš Slanina et al.
Journal of the American Chemical Society, 139(42), 15168-15175 (2017-10-19)
A detailed investigation of the photophysical parameters and photochemical reactivity of meso-methyl BODIPY photoremovable protecting groups was accomplished through systematic variation of the leaving group (LG) and core substituents as well as substitutions at boron. Efficiencies of the LG release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service