Skip to Content
Merck
All Photos(1)

Key Documents

907340

Sigma-Aldrich

H-L-Photo-Phe-OH

≥98%

Synonym(s):

(S)-2-Amino-3-(4-(3-(trifluoromethyl)-3H-diazirin-3-yl)phenyl)propanoic acid, 4-(Trifluoromethyldiazirin)-L-phenylalanine, Diazirine amino acid, H-Tdf-OH, Photo-Phe, Photo-crosslinking amino acid, Photoprobe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10F3N3O2
CAS Number:
Molecular Weight:
273.21
MDL number:
UNSPSC Code:
12352101

Assay

≥98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

FC(F)(F)C2(N=N2)c1ccc(cc1)CC(N)C(=O)O

InChI

1S/C11H10F3N3O2/c12-11(13,14)10(16-17-10)7-3-1-6(2-4-7)5-8(15)9(18)19/h1-4,8H,5,15H2,(H,18,19)

InChI key

HRGXDARRSCSGOG-UHFFFAOYSA-N

Application

H-L-Photo-Phe-OH is a diazirine-containing phenylalanine amino acid and multifunctional
photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (∼360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An Fmoc-protected version is also available as 907294.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Baldini et al.
Biochemistry, 27(20), 7951-7959 (1988-10-04)
The Boc-protected derivative of a photoactivatable, carbene-generating analogue of phenylalanine, L-4'-[3-(trifluoromethyl)-3H-diazirin-3-yl]phenylalanine [(Tmd)Phe], was used to acylate 5'-O-phosphorylcytidylyl(3'-5')adenosine (pCpA). A diacyl species was isolated which upon successive treatments with trifluoroacetic acid and 0.01 M HCl yielded a 1:1 mixture of 2'(3')-O-(Tmd)phenylalanyl-pCpA
A genetically encoded diazirine photocrosslinker in Escherichia coli.
Eric M Tippmann et al.
Chembiochem : a European journal of chemical biology, 8(18), 2210-2214 (2007-11-15)
Nobumasa Hino et al.
Journal of molecular biology, 406(2), 343-353 (2010-12-28)
Cell signaling pathways are essentially organized through the distribution of various types of binding domains in signaling proteins, with each domain binding to specific target molecules. Although identification of these targets is crucial for mapping the pathways, affinity-based or copurification
Lei Wang et al.
Molecules (Basel, Switzerland), 18(7), 8393-8401 (2013-07-19)
Photoaffinity labeling is a reliable analytical method for biological functional analysis. Three major photophores--aryl azide, benzophenone and trifluoromethyldiazirine--are utilized in analysis. Photophore-bearing L-phenylalanine derivatives, which are used for biological functional analysis, were inoculated into a Klebsiella sp. isolated from the
Lei Wang et al.
Bioscience, biotechnology, and biochemistry, 78(7), 1129-1134 (2014-09-18)
In this paper we report here a hydrogen/deuterium exchange (H/D exchange) of cross-linkable α-amino acid derivatives with deuterated trifluoromethanesulfonic acid (TfOD). H/D exchange with TfOD was easily applied to o-catechol containing phenylalanine (DOPA) within an hour. A partial H/D exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service