Skip to Content
Merck
All Photos(3)

Key Documents

905356

Sigma-Aldrich

Rh2(R-PTAD)4

Synonym(s):

Tetrakis[(R)-(-)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H80N4O16Rh2
CAS Number:
Molecular Weight:
1559.32
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder or crystals

mp

>300 °C

SMILES string

[Rh]([Rh](OC(=O)[C@H](N%19C(=O)c%20c(cccc%20)C%19=O)C%17%16C[C@@H]%18C[C@@H](C[C@@H](C%18)C%17)C%16)OC(=O)[C@H](N%14C(=O)c%15c(cccc%15)C%14=O)C%12%11C[C@@H]%13C[C@@H](C[C@@H](C%13)C%12)C%11)(OC(=O)[C@H](N9C(=O)c%10c(cccc%10)C9=O)C76C[C@@H]8C[C@@H](C[C@@H]

InChI

1S/4C20H21NO4.2Rh/c4*22-17-14-3-1-2-4-15(14)18(23)21(17)16(19(24)25)20-8-11-5-12(9-20)7-13(6-11)10-20;;/h4*1-4,11-13,16H,5-10H2,(H,24,25);;/q;;;;2*+2/p-4/t4*11-,12+,13-,16-,20?;;/m0000../s1

InChI key

SGEDWOHAUXKUGM-VMLWFVNWSA-J

Application

This chiral Rh(II) dimer was developed in the Davies lab to perform asymmetric carbene and nitrene reactions (C-H insertion, cyclopropanation, aziridination, C-H amination) with high regio- and stereo-control.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristian Soldi et al.
Journal of the American Chemical Society, 136(43), 15142-15145 (2014-10-14)
The first asymmetric insertion reactions of donor-donor carbenoids, i.e., those with no pendant electron-withdrawing groups, are reported. This process enables the synthesis of densely substituted benzodihydrofurans with high levels of enantio- and diastereoselectivity. Preliminary results show similar efficiency in the
Andrew DeAngelis et al.
Journal of the American Chemical Society, 133(6), 1650-1653 (2011-01-27)
A catalytic, enantioselective method for the C-H functionalization of indoles by diazo compounds has been achieved. With catalytic amounts of Rh(2)(S-NTTL)(4), the putative Rh-carbene intermediates from α-alkyl-α-diazoesters react with indoles at C(3) to provide α-alkyl-α-indolylacetates in high yield and enantioselectivity.
Brett D Schwartz et al.
Journal of the American Chemical Society, 131(23), 8329-8332 (2009-05-19)
The total synthesis of (-)-5-epi-vibsanin E (2) has been achieved in 18 steps. The synthesis combines the rhodium-catalyzed [4 + 3] cycloaddition between a vinylcarbenoid and a diene to rapidly generate the tricyclic core with an effective end game strategy
Ravisekhara P Reddy et al.
Organic letters, 8(16), 3437-3440 (2006-07-28)
[reaction: see text] The dirhodium tetracarboxylate, Rh2(S-PTAD)4, derived from adamantylglycine, is a very effective chiral catalyst for carbenoid reactions. High asymmetric induction was obtained in Rh2(S-PTAD)4-catalyzed intramolecular C-H insertion (94% ee), intermolecular cyclopropanation (99% ee), and intermolecular C-H insertion (92%
Asymmetric synthesis of tropanes by rhodium-catalyzed [4 + 3] cycloaddition.
Ravisekhara P Reddy et al.
Journal of the American Chemical Society, 129(34), 10312-10313 (2007-08-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service