Skip to Content
Merck
All Photos(2)

Key Documents

904864

Sigma-Aldrich

(S,R,S)-AHPC-PEG6-butyl azide

≥95%

Synonym(s):

(2S,4R)-1-((S)-27-Azido-2-(tert-butyl)-4-oxo-6,9,12,15,18,21-hexaoxa-3-azaheptacosanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, (S,R,S)-AHPC-2-2-2-2-2-2-6-N3, Crosslinker−E3 Ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader, VH032 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H63N7O10S
Molecular Weight:
834.03
UNSPSC Code:
51171641
NACRES:
NA.22

ligand

VH032

Assay

≥95%

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: ligand-linker conjugate

functional group

azide

storage temp.

2-8°C

SMILES string

O=C(N[C@H](C(N1[C@H](C(NCC2=CC=C(C3=C(C)N=CS3)C=C2)=O)C[C@@H](O)C1)=O)C(C)(C)C)COCCOCCOCCOCCOCCOCCCCCCN=[N+]=[N-]

Application

Protein degrader builiding block (S,R,S)-AHPC-PEG6-butyl azide enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a von Hippel–Lindau (VHL)-recruiting ligand, a linker with both hydrophobic and hydrophilic moieties, and a pendant azide for click chemistry with a target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant azide group, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service