Skip to Content
Merck
All Photos(1)

Key Documents

90460

Sigma-Aldrich

Triethylenetetramine

≥97.0% (T)

Synonym(s):

TETA, DEH 24, EPH 925

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2
CAS Number:
Molecular Weight:
146.23
Beilstein:
605448
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~5 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥97.0% (T)

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

266-267 °C (lit.)

mp

12 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCNCCNCCN

InChI

1S/C6H18N4/c7-1-3-9-5-6-10-4-2-8/h9-10H,1-8H2

InChI key

VILCJCGEZXAXTO-UHFFFAOYSA-N

Gene Information

human ... TERT(7015)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Triethylenetetramine has been used as an additive to enhance the peak resolution ability of capillary zone electrophoresis (CZE) running buffer system to separate and quantitate the monoclonal antibodies by the CZE method. It may be used for the amination of polyacrylonitrile fibers to form novel fiber catalysts for Knoevenagel condensation in aqueous media. TETA also acts as a copper (II)-selective chelator. It may also be used as a growth-orientator in the formation of 1D zinc sulfide nanoarchitectures.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

264.2 °F

Flash Point(C)

129 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and reusable amine-functionalized polyacrylonitrile fiber catalysts for Knoevenagel condensation in water.
Li G
Green Chemistry, 14(8), 2234-2242 (2012)
Lars Roll et al.
Frontiers in cellular neuroscience, 14, 102-102 (2020-06-09)
The mammalian central nervous system (CNS) is characterized by a severely limited regeneration capacity. Comparison with lower species like amphibians, which are able to restore even complex tissues after damage, indicates the presence of an inhibitory environment that restricts the
Triethylenetetramine (TETA)-assisted synthesis, dynamic growth mechanism, and photoluminescence properties of radial single-crystalline ZnS nanowire bundles.
Liu J
Journal of Crystal Growth, 311(5), 1423-1429 (2009)
Development and validation of a rapid capillary zone electrophoresis method for determining charge variants of mAb.
Shi Y
Journal of Chromatography. B, Biomedical Applications, 906, 63-68 (2012)
Metabolomic analysis of rat serum in streptozotocin-induced diabetes and after treatment with oral triethylenetetramine (TETA).
Ugarte M
Genome Medicine, 4(4), 35-35 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service