Skip to Content
Merck
All Photos(1)

Documents

901756

Sigma-Aldrich

Low endotoxin gelatin from porcine skin

gel strength 240-360 (Bloom), <10 EU/g Endotoxin

Synonym(s):

beMatrix gelatin LS-H

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116130
NACRES:
NA.23

form

powder

impurities

<10 EU/g Endotoxin
<300 g Total viable aerobic count

loss

<15% loss on drying

color

white to pale yellow

pH

5.0-6.5

viscosity

4.0-6.5 mPa.s

anion traces

sulfite (SO32-): <1.5 mg/20g

cation traces

As: <1 ppm
Hg: <0.1 ppm
heavy metals: <20 ppm

Looking for similar products? Visit Product Comparison Guide

Application

Gelatin has received significant attention in the biomedical field due to its inherent bioactivity. Low endotoxin gelatin from porcine skin, gel strength 300 bloom is a high bloom (high molecular weight) gelatin featuring less than 10 endotoxin units (EU) per gram. Endotoxins primarily consist of lipopolysaccharides from the outer membrane of gram-negative bacteria. Once in the bloodstream, endotoxins are capable of eliciting a strong immune response, potentially leading to fever, shock, a drop in blood pressure, and even potentially death. In addition to this immune response, several reports suggest endotoxins may also damage various types of cells. To avoid adverse effects, raw materials should contain the lowest level of endotoxins possible when used in biomedical or regenerative medicine research applications. This low endotoxin gelatin is derived from alkaline-treated porcine skin and has a variety of potential applications in the fields of regenerative medicine, medical devices, and tissue engineering research.

Legal Information

beMatrix is a trademark of Nitta Gelatin

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K W Brunson et al.
Journal of supramolecular structure, 9(2), 231-242 (1978-01-01)
Chinese hamster ovary (CHO . K1 . PRO) cell growth was inhibited by addition of a gram-negative bacterial lipopolysaccharide (LPS) to the cell culture medium. Growth inhibition began after three or four days of incubation, was dose-dependent up to a
J M Harlan et al.
Laboratory investigation; a journal of technical methods and pathology, 48(3), 269-274 (1983-03-01)
Lipopolysaccharide (LPS) produced time- and dose-dependent bovine endothelial cell injury in vitro that was manifested initially by cell detachment from culture substrate with subsequent cell lysis. Bovine endothelial cell injury was observed with LPS derived from Salmonella minnesota R595, a
J G Brock-Utne et al.
Anaesthesia and intensive care, 17(1), 49-55 (1989-02-01)
Endotoxins (lipopolysaccharides, LPS) are potent bacterial poisons always present within the intestines in considerable amounts. Several pathophysiological conditions such as hypovolaemia, hypoxia, intestinal ischaemia, burns and radiation lead to a breakdown in the barrier and depending upon the extent of
E J Ziegler et al.
The New England journal of medicine, 307(20), 1225-1230 (1982-11-11)
In an effort to decrease deaths from gram-negative bacteremia and endotoxin shock, we treated bacteremic patients with human antiserum to endotoxin (lipopolysaccharide) core. Antiserum was prepared by vaccinating healthy men with heat-killed Escherichia coli J5; this mutant lacks lipopolysaccharide oligosaccharide
J Ongrádi et al.
Folia microbiologica, 29(6), 450-454 (1984-01-01)
Cytotoxicity of a mixed pyrogen preparation and its components as well as native and radiodetoxified lipopolysacharides (LPS) was determined with established HEp-2 cell cultures and by measuring plating efficiency. This proved to be more sensitive to the damaging effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service