Skip to Content
Merck
All Photos(1)

Key Documents

900724

Sigma-Aldrich

Molybdenum disulfide

greener alternative

dispersion, 0.1-0.5 mg/mL in H2O

Synonym(s):

2D Dispersion, MoS2 Dispersion nonionic surfactant, MoS2 Dispersion with Pluronic F87, Moly sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoS2
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.23

form

dispersion

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.1-0.5 mg/mL in H2O

particle size

≤500 nm

bp

100 °C (water)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

[Mo](=S)=S

InChI

1S/Mo.2S

InChI key

CWQXQMHSOZUFJS-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

The two dimensional layered inorganic nanomaterials have drawn significant attention recently owing to their thickness dependent physical and chemical properties. They find applications in photocatalysis, photovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc, transition metal dichalcogenides represent an important class of inorganic 2D materials having the aforementioned applications.

Legal Information

Sold under Material Transfer Agreement with Mark Hersam group at Northwestern University.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar JL, et al.
The Journal of Physical Chemistry C, 118, 3847-3863 (2014)
Nikhita D Mansukhani et al.
Small (Weinheim an der Bergstrasse, Germany), 12(3), 294-300 (2015-12-01)
Conditions for the dispersion of molybdenum disulfide (MoS2) in aqueous solution at concentrations up to 0.12 mg mL(-1) using a range of nonionic, biocompatible block copolymers (i.e., Pluronics and Tetronics) are identified. Furthermore, the optimal Pluronic dispersant for MoS2 is
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service