Skip to Content
Merck
All Photos(1)

Key Documents

900534

Sigma-Aldrich

(t-Bu)PhCPhos Pd G3

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H46N3O3PPdS
CAS Number:
Molecular Weight:
774.26
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

Application

As the Pd G3 complex, this ligand from the Buchwald group offers best-in-class performance in the arylation of sterically hindered primary amines.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Ruiz-Castillo et al.
Journal of the American Chemical Society, 137(8), 3085-3092 (2015-02-05)
We report the Pd-catalyzed arylation of very hindered α,α,α-trisubstituted primary amines. Kinetics-based mechanistic analysis and rational design have led to the development of two biarylphosphine ligands that allow the transformation to proceed with excellent efficiency. The process was effective in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service