Skip to Content
Merck
All Photos(1)

Key Documents

900526

Sigma-Aldrich

Doped graphene

boron-doped, avg. no. of layers, 1 ‑ 5

Synonym(s):

B-doped graphene, BG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

Product Name

Boron-doped graphene, avg. no. of layers, 1 ‑ 5

description

Oxygen: <7.5%

Quality Level

Assay

≥95%

form

powder

feature

avg. no. of layers 1 ‑ 5

surface area

>500 m2/g

cation traces

B: 2.0-4.0%
C: 85-95%

General description

  • Typical thickness: 1-5 layers.
  • Typical size : 0.5-5 μm.

Application

  • Electro catalyst.
  • Field-effect transistors.
  • Sensors.
  • Lithium ion batteries.
  • Supercapacitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron-doped graphene as high-performance electrocatalyst for the simultaneously electrochemical determination of hydroquinone and catechol.
Zhang, et al.
Electrochimica Acta, 156, 228-234 (2015)
Zhong-Shuai Wu et al.
ACS nano, 5(7), 5463-5471 (2011-06-24)
One great challenge in the development of lithium ion batteries is to simultaneously achieve high power and large energy capacity at fast charge and discharge rates for several minutes to seconds. Here we show that nitrogen- or boron-doped graphene can
First-Principles Study of Nitrogen-, Boron-Doped Graphene and Co-Doped Graphene as the Potential Catalysts in Nonaqueous LiO2 Batteries.
Jiang HR, et al.
The Journal of Physical Chemistry C, 120(12), 6612-6618 (2016)
Mingyang Xing et al.
Scientific reports, 4, 5493-5493 (2014-07-01)
TiO2/graphene (TiO2-x/GR) composites, which are Ti(3+) self-doped TiO2 nanorods decorated on boron doped graphene sheets, were synthesized via a simple one-step hydrothermal method using low-cost NaBH4 as both a reducing agent and a boron dopant on graphene. The resulting TiO2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service