Skip to Content
Merck
All Photos(1)

Key Documents

87212

Sigma-Aldrich

Trimethyl-tetradecylammonium chloride

≥98.0% (AT)

Synonym(s):

Tetradecyltrimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Cl)(CH3)3
CAS Number:
Molecular Weight:
291.94
Beilstein:
3916572
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

powder

mp

~250 °C (dec.)

SMILES string

[Cl-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CEYYIKYYFSTQRU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dulan B Gunasekara et al.
Electrophoresis, 32(8), 832-837 (2011-03-26)
The combination of microchip electrophoresis with amperometric detection leads to a number of analytical challenges that are associated with isolating the detector from the high voltages used for the separation. While methods such as end-channel alignment and the use of
Dawei Fan et al.
Journal of colloid and interface science, 333(2), 757-763 (2009-02-24)
Phase stability of two nanometer-scale Keplerate-type polyoxomolybdates, (NH(4))(42)[Mo(VI)(72)Mo(V)(60)O(372)(CH(3)COO)(30)(H(2)O)(72)]300H(2)O10CH(3)COONH(4) ({Mo(132)}) and Mo(VI)(72)Fe(III)(30)O(252)L(102)ca. 180H(2)O with L=H(2)O/CH(3)COO(-)/Mo(2)O(n-)(8/9) ({Mo(72)Fe(30)}), can be easily achieved by controlling the concentration of a cationic surfactant, tetradecyltrimethylammonium bromide (TTABr), in aqueous solution. Precipitates and floccules were observed when the
Yuri Borodko et al.
Langmuir : the ACS journal of surfaces and colloids, 25(12), 6665-6671 (2009-04-29)
The vibrational spectra of platinum nanoparticles (12 nm) capped with tetradecyltrimethylammonium bromide, C(14)TAB, were investigated by Fourier transform infrared (FTIR) spectroscopy. We have shown that the thermal decay of Pt-C(14)TAB nanoparticles in N(2), H(2), and O(2) atmospheres leads to the
Aline Delbos et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 84(1 Pt 1), 011404-011404 (2011-08-27)
We investigate experimentally the behavior of liquid foams pumped at a given flow rate through a single pore, in the situation where the pore diameter is smaller than the bubble diameter. Results reveal that foam invasion can be observed only
Hayat M Marafie et al.
Talanta, 91, 26-32 (2012-03-01)
Two conventional plastic membrane electrodes that are selective for the tetradecyltrimethylammonium cation (TTA) have been prepared. The ion exchangers of these sensors were the ion associate, TTA-PT, and the ion aggregate, TTA-PSS, where PT and PSS are phosphotungstate and polystyrene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service