Skip to Content
Merck
All Photos(1)

Key Documents

855286

Sigma-Aldrich

5-Aminouracil

98%

Synonym(s):

5-Amino-2,4-dihydroxypyrimidine, 5-Amino-2,4-pyrimidinediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Beilstein:
127250
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

NC1=CNC(=O)NC1=O

InChI

1S/C4H5N3O2/c5-2-1-6-4(9)7-3(2)8/h1H,5H2,(H2,6,7,8,9)

InChI key

BISHACNKZIBDFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulina Spisz et al.
International journal of molecular sciences, 21(17) (2020-09-05)
Hypoxia-a hallmark of solid tumors-dramatically impairs radiotherapy, one of the most common anticancer modalities. The adverse effect of the low-oxygen state can be eliminated by the concomitant use of a hypoxic cell radiosensitizer. In the present paper, we show that
F Cortés et al.
Mutation research, 247(1), 147-151 (1991-03-01)
Poly-D-lysine has been reported to induce a triggering of mitosis in plant cells due to a selective stimulatory effect on cells arrested in G2. Root-tip cells of Allium cepa L. were first exposed to maleic hydrazide (MH) early in the
Recovery from the 5-AU induced blockage in interphase: evidence for differential recovery.
D Davidson
Cytologia, 47(3-4), 545-553 (1982-12-01)
M H Navarrete et al.
Experimental cell research, 148(2), 273-280 (1983-10-15)
Meristematic cells from Allium cepa L roots can attain a steady-state of growth at both 15 and 25 degrees C in the presence of drugs, hydroxyurea and 5-amino-uracil, which reduce the rate of DNA synthesis. These drugs, at used concentrations
K K Upadhyay et al.
Organic & biomolecular chemistry, 8(21), 4892-4897 (2010-09-08)
A new fluorescent probe (5-[(4-diethylamino-2-hydroxy-benzylidene)-amino]-1H-pyrimidine-2, 4-dione) (Receptor 1) has been synthesized by the Schiff base condensation of 5-aminouracil with 4-(diethylamino)salicylaldehyde. The receptor 1 exhibits high selectively for Al(3+) in DMSO as well as in aqueous solution even in the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service