Skip to Content
Merck
All Photos(1)

Key Documents

85419

Sigma-Aldrich

Silicone oil AR 200

viscosity ~200 mPa.s,   neat(25 °C)

Synonym(s):

Polyphenyl-methylsiloxane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
Pricing and availability is not currently available.

form

oil

Quality Level

refractive index

n20/D 1.453

viscosity

~200 mPa.s, neat(25 °C)

density

1.05 g/mL at 20 °C

General description

Silicone oil AR 200 is a polydimethylsiloxane with phenyl groups typically used as a heat transfer or pressure transfer fluid.

Application

Silicone oil AR 200 can be used as:
  • A chemical modifier to modify thermosetting phenolic resin to prepare adhesive.
  • A dopant for 4′-n-octyl-4-cyano-biphenyl liquid crystal matrix.

Features and Benefits

Especially good thermostability

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

545.0 °F

Flash Point(C)

285 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kriegs et al.
The Journal of chemical physics, 124(10), 104901-104901 (2006-03-18)
In some polymers, in addition to the usual structural alpha relaxation, a slower alpha' relaxation is observed with a non-Arrhenius temperature dependence. In order to understand better the molecular origin of this alpha' relaxation in poly(methylphenylsiloxane) (PMPS) we have studied
C Eckardt et al.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde, 201(3), 133-139 (1990-01-01)
Different methylphenylsilicone (MPS) and fluorosilicone (FS) oils having a higher specific gravity than water were investigated as vitreous replacements. Their intraocular tolerance in vitrectomized aphakic rabbit eyes was evaluated for periods of up to 4 months and compared to that
Experience with a silicone resin (polymethylphenylsiloxane) hard contact lens.
H Magatani et al.
The CLAO journal : official publication of the Contact Lens Association of Ophthalmologists, Inc, 9(3), 261-266 (1983-07-01)
O Gyllenhaal et al.
Journal of chromatography, 516(2), 415-426 (1990-09-21)
The supercritical fluid chromatography of intact aliphatic amines with different columns is described. One group of amines was based on N,N-dimethyl-n-octylamine and related primary and secondary amines, and the other on the amino alcohol metoprolol and several of its analogues.
G B Chen et al.
Se pu = Chinese journal of chromatography, 18(4), 343-345 (2003-01-25)
An insoluble beta-cyclodextrin polymer cross-linked with epichlorohydrin was prepared. beta-Cyclodextrin polymer (0.26 g) was treated for 0.5 h by a ultrasonator in the mixed solvent (10 mL) of isopropyl ether, methylene dichloride and benzene with a volume ratio 2:1:2, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service