Skip to Content
Merck
All Photos(2)

Key Documents

806889

Sigma-Aldrich

Silica

mesoporous, 1 μm particle size, pore size ~4 nm

Synonym(s):

Porous Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9% trace metals basis

Quality Level

form

mesoporous
powder

surface area

300-400 m2/g

particle size

1 μm

pore size

0.2-0.4 cm3/g pore volume
~4 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mesoporous silicas are inorganic materials having porous structures and find applications in chromatography, catalysis, drug delivery and imaging.[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang Y
Advanced Materials, 273, 576-576 (2015)
Si-Han Wu et al.
Chemical Society reviews, 42(9), 3862-3875 (2013-02-14)
Good control of the morphology, particle size, uniformity and dispersity of mesoporous silica nanoparticles (MSNs) is of increasing importance to their use in catalyst, adsorption, polymer filler, optical devices, bio-imaging, drug delivery, and biomedical applications. This review discusses different synthesis
Isabel Sierra et al.
Journal of chromatography. A, 1363, 27-40 (2014-07-13)
Recent advances in the development of new materials are having a major impact on analytical chemistry. For example, the unique properties of ordered mesoporous silicas (OMSs) have been shown to enhance the analytical performance of many existing techniques or allow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service