802565
Aquivion® D79-25BS
liquid, dispersion, 25% in water, PFSA eq. wt. 790 g/mole SO3H, contains CF3 polymer chain ends as stabilizer
Synonym(s):
Aquivion® SO3H-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C4HF7O4S .C2F4)x
UNSPSC Code:
26111700
NACRES:
NA.23
Quality Level
form
dispersion
liquid
eq. wt.
790±20g/mol (SO3H)
contains
CF3 polymer chain ends as stabilizer
composition
25% in water
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
25±1 % (w/w)
density
1.14 g/mL
greener alternative category
Looking for similar products? Visit Product Comparison Guide
General description
Aquivion® PFSA dispersions are derived from a short-side-chain (SSC) copolymer of Tetrafluoroethylene and Sulfonyl Fluoride Vinyl Ether (SFVE), CF2=CF2-O-(CF2)2-SO2F, industrially produced by Syensqo Group. The resulting perfluoropolymer features functional groups in their sulfonic acid form (SO3H). Aquivion® D79-25BS is a perfluorosulfonic acid (PFSA) ionomer dispersion with an Equivalent Weight (EW) of 790 g/eq. The EW of an ionomer represents the weight of the polymer required to provide 1 mole of exchangeable protons, inversely related to the ion exchange capacity (IEC). This dispersion is chemically stabilized, with a significant proportion of polymer end groups converted to an inert trifluoromethyl (CF3) configuration, enhancing the polymer′s lifetime. Aquivion® D79-25BS contains a polymer concentration of 25 wt% in a solvent system that is over 99% water and free of ethers.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for use as Proton Exchange Membrane (PEM) Fuel Cells. Click here for more information.
Application
The superacidity of Aquivion® PFSA resins, along with their high functional group concentration and electrochemical stability, makes them essential components in heterogeneous catalysts. Because of their excellent proton conductivity at low water uptake, these resins are used frequently in research as ionomer binders within the catalyst layers of proton exchange membranes (PEM) for fuel cells and water electrolyzers. Additionally, these resins are utilized in the fabrication of separator membranes (PEMs), either as stand-alone materials or reinforced with PTFE. They are also applied to the surfaces of specialty fluoropolymers to enhance wettability. Aquivion® D79-25BS, characterized by its short side chain and moderate equivalent weight, is a promising ionomer for achieving high catalyst activity. Ionomers with lower equivalent weights, such as D79-25BS, exhibit greater proton conductivities in electrochemical tests. This makes D79-25BS particularly suitable for use in PEM fuel cells and water electrolyzers. Additionally, it has proven to be an effective ionomer for heterogeneous catalysts in carbon dioxide and carbon monoxide electrochemistry. Researchers have successfully combined D79-25BS with platinum group catalysts and transition metal catalysts for various chemical reactions, owing to the ionomer’s resistance to highly aggressive chemical environments, including low pH, strong oxidants, and strong reducing agents.
Features and Benefits
- Very high concentration of functional groups for strong proton (or Na+, K+, Li+, etc.) conductivity
- High softening point for high temperature operation well above 80 ºC •
- Strong water retention for better fuel cell operation at low-humidity conditions for membranes or catalyst inks that are utilizing this dispersion material
Legal Information
Aquivion is a registered trademark of Syensqo Group
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service