798320
2-Bromo-6-(trimethylsilyl)phenyl triflate
95%
Synonym(s):
Garg 3-Bromobenzyne Precursor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H12BrF3O3SSi
CAS Number:
Molecular Weight:
377.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n/D 1.492
density
1.482 g/L at 25 °C
functional group
bromo
fluoro
triflate
storage temp.
2-8°C
SMILES string
O=S(OC1=C([Si](C)(C)C)C=CC=C1Br)(C(F)(F)F)=O
InChI
1S/C10H12BrF3O3SSi/c1-19(2,3)8-6-4-5-7(11)9(8)17-18(15,16)10(12,13)14/h4-6H,1-3H3
InChI key
ZNKAHJCGEPVEBA-UHFFFAOYSA-N
Application
Synchronous Ar-F and Ar-Sn Bond Formation Through Fluorostannylation of Arynes
Continuous-Flow Synthesis of Trimethylsilylphenyl Perfluorosulfonate Benzyne Precursors
Continuous-Flow Synthesis of Trimethylsilylphenyl Perfluorosulfonate Benzyne Precursors
Bromosilyltriflate has been shown to be a useful precursor for the generation of a bromobenzyne intermediate. The bromobenzyne intermediate can be generated in the presence of cesium or potassium fluoride and can be trapped with various dipolariphiles.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service