Skip to Content
Merck
All Photos(1)

Key Documents

794139

Sigma-Aldrich

5 -Cyclopentadienyl)[N,N-dimethyl-Ν′-(2-pyridinylmethylidene)propane-1,3-diamine]ruthenium hexafluorophosphate

Synonym(s):

Herzon Reductive Hydration Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22F6N3PRu
CAS Number:
Molecular Weight:
502.40
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: ruthenium
reagent type: catalyst

mp

>180 °C

SMILES string

CN(C)CCC/N=C/C1=NC=CC=C1.F[P-](F)(F)(F)(F)F.[C]2[C][C][C][C]2.[Ru+]

InChI

1S/C11H17N3.C5H5.F6P.Ru/c1-14(2)9-5-7-12-10-11-6-3-4-8-13-11;1-2-4-5-3-1;1-7(2,3,4,5)6;/h3-4,6,8,10H,5,7,9H2,1-2H3;1-5H;;/q;;-1;+1/b12-10+;;;

InChI key

KUSUXOOKLDLSSI-AKMNYRKMSA-N

General description

5 -Cyclopentadienyl)[N,N-dimethyl-N′-(2-pyridinylmethylidene)propane-1,3-diamine]ruthenium hexafluorophosphate is also referred to as Herzon reductive hydration catalyst. Herzon reductive hydration catalyst is a half-sandwich ruthenium complex. It is a tridentate nitrogen-based ligand containing a hemilabile 3-(dimethylamino)propyl group. It participates in the conversion of terminal alkynes to linear alcohols.

Application

Catalyst mediates the transformation of alkynes to alcohols through an anti-Markovnikov addition. This reaction occurs at room temperature with the use of formic acid to assist in the reduction of the intermediate aldehyde to the corresponding alcohol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingshuo Zeng et al.
Journal of the American Chemical Society, 136(19), 7058-7067 (2014-05-03)
The conversion of terminal alkynes to functionalized products by the direct addition of heteroatom-based nucleophiles is an important aim in catalysis. We report the design, synthesis, and mechanistic studies of the half-sandwich ruthenium complex 12, which is a highly active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service