Skip to Content
Merck
All Photos(1)

Key Documents

792586

Sigma-Aldrich

tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO4
CAS Number:
Molecular Weight:
241.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

refractive index

n20/D 1.474

density

1.052 g/mL at 25 °C

functional group

ester

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(N(C1)CC1=CC(OCC)=O)=O

InChI

1S/C12H19NO4/c1-5-16-10(14)6-9-7-13(8-9)11(15)17-12(2,3)4/h6H,5,7-8H2,1-4H3

InChI key

WYWJZDFQQJTRDD-UHFFFAOYSA-N

Application

Building block used as a precursor to the synthesis of thia and oxa-azaspiro[3.4]octanes. As reported in the Carreira lab, the product can readily undergo [3+2] cycloadditions with dipolariphiles to generate a series of small-ring spirocycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service