Skip to Content
Merck
All Photos(3)

Key Documents

790907

Sigma-Aldrich

TmPyPB

98% (HPLC)

Synonym(s):

1,3,5-Tri(m-pyridin-3-ylphenyl)benzene, 1,3,5-Tris(3-pyridyl-3-phenyl)benzene, 3,3′-[5′-[3-(3-pyridinyl)phenyl][1,1′:3′,1′′-terphenyl]-3,3′′-diyl]bispyridine, Tm3PyPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H27N3
CAS Number:
Molecular Weight:
537.65
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98% (HPLC)

form

powder

mp

195-200 °C

SMILES string

C1(C2=CC=CC(C3=CN=CC=C3)=C2)=CC(C4=CC=CC(C5=CN=CC=C5)=C4)=CC(C6=CC=CC(C7=CN=CC=C7)=C6)=C1

InChI

1S/C39H27N3/c1-7-28(34-13-4-16-40-25-34)19-31(10-1)37-22-38(32-11-2-8-29(20-32)35-14-5-17-41-26-35)24-39(23-37)33-12-3-9-30(21-33)36-15-6-18-42-27-36/h1-27H

InChI key

CINYXYWQPZSTOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Electron-transport and hole/exciton-blocking materail with high electron mobility (10-4-10-3 cm2 V-1 s-1) and high triplet energy level (2.75 eV) for highly efficient phosphorescent OLEDs application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-property relationship of pyridine-containing triphenyl benzene electron-transport materials for highly efficient blue phosphorescent OLEDs
Su, Shi-Jian; Takahashi, Yasuyuki; et al.
Advances in Functional Materials, 19(8), 1260-1267 (2009)
Pyridine-containing triphenylbenzene derivatives with high electron mobility for highly efficient phosphorescent OLEDs
Su, Shi-Jian; Chiba, Takayuki; et al.
Advanced Materials, 20(11), 2125-2130 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service