Skip to Content
Merck
All Photos(1)

Key Documents

790192

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

stabilized with octadecylamine ligands, fluorescence λem 620 nm, solid

Synonym(s):

Artificial atoms, Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

form

solid

fluorescence

λem 620 nm

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

CdSe/ZnS core/shell nanocrystals in solid form are highly purified quantum dots. They can be dispersed in most non-polar organic solvents such as toluene, chloroform, hexane, etc. With very low organic impurity, these quantum dots are specifically designed as emitters for optoelectronic applications such as display and solid state lighting.

Protocol to make QD solutions (dispersing QDs into solutions):

1. Weigh desired amount of the quantum dot powder and put it in a glass vial;
2. Add desired amount of the solvent, i.e. toluene, chloroform, hexane, etc. (mostly non-polar organic solvents) in the vial and shake the vial;
3. If there is any solid left in the vial after shaking, sonicate the vial for 30~60 seconds to get the quantum dot powder well dispersed. If required dilute the solution further.

Material has a shelf life of around 3 years if stored properly. Store at room temperature (4-25 °C); do not freeze. Should not be exposed to extreme temperatures.

Legal Information

Product of Ocean Nanotech, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Oral

Target Organs

Kidney,Bone, Liver,Gastrointestinal tract,Immune system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service