Skip to Content
Merck
All Photos(2)

Documents

778583

Sigma-Aldrich

Azidobenzene solution

greener alternative

~0.5 M in 2-methyltetrahydrofuran, ≥95.0% (HPLC)

Synonym(s):

Phenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
Beilstein:
742248
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

~0.5 M in 2-methyltetrahydrofuran

impurities

≤2.0% water

greener alternative category

storage temp.

−20°C

SMILES string

[N-]=[N+]=Nc1ccccc1

InChI

1S/C6H5N3/c7-9-8-6-4-2-1-3-5-6/h1-5H

InChI key

CTRLRINCMYICJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azidobenzene is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.
Azidobenzene solution can undergo click reaction with alkyne-functionalized oligomers to form O-ester-functionalized o-phenylenes, a group of aromatic foldamers with a good folding tendency in most solvents.

Application

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Viel et al.
Langmuir : the ACS journal of surfaces and colloids, 29(6), 2075-2082 (2013-01-16)
Covalent immobilization of unmodified biological materials as proteins has been performed through a one-step and soft method. This process is based on a polyazidophenylene layer derived from the electroreduction of the parent salt 4-azidobenzenediazonium tetrafluoborate on gold substrates. The wavelength
Laëtitia Renaudie et al.
Biomacromolecules, 8(2), 679-685 (2007-02-13)
Grafting of a new carbohydrate UV-reactive molecule, an azidophenyl lactamine (AzPhLac), was achieved on fibers of three different diameters: 12, 18, and 32 microm. Adsorption of AzPhLac on fibers was obtained by using the dip-coating method in solution. The effect
C Malaveille et al.
Cancer research, 42(4), 1446-1453 (1982-04-01)
3-Methyl-1-phenyltriazene and a series of ring-substituted derivatives (4-methylphenyl, 4-chlorophenyl, and 2,4,6-trichlorophenyl), structurally related benzenediazonium fluoborates and phenyl azides, as well as the recently isolated [1-methyl-3-(2,4,6-trichlorophenyl)-2-triazeno]methyl-beta-D-glucopyranoside uronic acid, were studied for their mutagenic activity in Salmonella typhimurium strains. Of these compounds
Solvent effects on the folding of o-phenylene oligomers
Vemuri GN, et al.
Organic & Biomolecular Chemistry, 15(4), 845-851 (2017)
Yutaka Sadakane et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(2), 209-218 (2006-03-04)
Photoaffinity labeling enables the direct probing of a target protein through a covalent bond between a ligand and its binding protein, and even a complex formed by weak interactions can be isolated by the method. The photochemical fishing approach accelerates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service