Skip to Content
Merck
All Photos(4)

Key Documents

775061

Sigma-Aldrich

Potassium bis(trimethylsilyl)amide solution

greener alternative

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

Hexamethyldisilazane potassium salt solution, Potassium hexamethyldisilazide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NK
CAS Number:
Molecular Weight:
199.48
Beilstein:
4006754
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M in 2-methyltetrahydrofuran

density

0.884 g/mL at 25 °C

greener alternative category

SMILES string

C[Si](C)(C)N([K])[Si](C)(C)C

InChI

1S/C6H18NSi2.K/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

IUBQJLUDMLPAGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with "Safer Solvents and Auxiliaries", "Use of Renewable Feedstocks" and "Inherently Safer Chemistry for Accident Prevention". Click here for more information.

Application

Strong organic base in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Nan Li et al.
Lab on a chip, 8(12), 2105-2112 (2008-11-22)
High-density live cell array serves as a valuable tool for the development of high-throughput immunophenotyping systems and cell-based biosensors. In this paper, we have, for the first time, demonstrated a simple fabrication process to form the hexamethyldisilazane (HMDS) and poly(ethylene
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six
Zhengtao Deng et al.
Nanoscale, 3(10), 4346-4351 (2011-09-15)
Here we report the colloidal synthesis of size-tunable SnS nanocrystals that have an unusual meta-stable cubic zinc-blende phase instead of the more stable layered orthorhombic phase. The single-crystalline zinc-blende SnS nanocrystals with sizes of 8 nm, 60 nm, and 700
Mario Janjetovic et al.
Chemical communications (Cambridge, England), 49(18), 1826-1828 (2013-01-30)
Sm(HMDS)(2) in n-hexane mediates fast cleavage of primary, secondary and tertiary alkyl fluorides in good to excellent yields. In n-hexane Sm(HMDS)(2) exhibits uniquely enhanced reductive ability towards the C-F bond compared to when using THF as solvent.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service