Skip to Content
Merck
All Photos(1)

Key Documents

769894

Sigma-Aldrich

Resomer® LG 824 S, Poly(L-lactide-co-glycolide)

ester terminated, Lactide: Glycolide 82:18, lactide:glycolide 82:12

Synonym(s):

Resomer® LG 824 S, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[C3H4O2]n[C2H2O2]mCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

solid

feed ratio

lactide:glycolide 82:12

viscosity

1.7-2.6 dL/g

mp

157-164 °C

transition temp

Tg 54-60 °C

storage temp.

2-8°C

General description

Resomer® LG 824 S, Poly(L-lactide-co-glycolide) (PLGA), is a semicrystalline, biocompatible, bioresorbable and biodegradable material with high porosity and high surface area. It can be processed by a variety of techniques such as electrospinning, electrospraying and wet spinning.

Application

PLGA can be used in the formation of a membrane for medical applications. It can also be used as a scaffold in tissue engineering.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (l-lactide-co-glycolide) biodegradable microfibers and electrospun nanofibers for nerve tissue engineering: an in vitro study
Bini TB, et al.
J. Mater. Sci., 41(19), 6453-6459 (2006)
In vitro biocompatibility of bioresorbable polymers: poly (L, DL-lactide) and poly (L-lactide-co-glycolide)
Ignatius AA and Claes LE
Biomaterials, 17(8), 831-839 (1996)
Poly (L-lactide-co-glycolide) microporous membranes for medical applications produced with the use of polyethylene glycol as a pore former
Krok M and Pamula E
Journal of Applied Polymer Science, 125(S2), E187-E199 (2012)
Electrospun poly (L-lactide-co-glycolide) biodegradable polymer nanofibre tubes for peripheral nerve regeneration
Bini TB, et al.
Nanotechnology, 15(11), 1459-1459 (2004)
Influences of tensile load on in vitro degradation of an electrospun poly (l-lactide-co-glycolide) scaffold
Li P, et al.
Acta Biomaterialia, 6(8), 2991-2996 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service