767719
Mytilus edulis foot protein-1
10 mg/mL (in 1% citric acid)
Synonym(s):
MEFP-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
Assay
≥90%
form
liquid
concentration
10 mg/mL (in 1% citric acid)
refractive index
n20/D 1.335
bp
100-102 °C/760 mmHg
density
1.008 g/mL at 25 °C
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
A solution of MEFP-1 for general applications such as formation of surface coatings, attaching particles or polymers to surfaces or adhesive compositions that are insensitive to water. A surface coating of MEFP-1 alters the surface to be highly hydrophilic and allows for attachment of other biological or non-biological components such as proteins, particles or polymers.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J H Waite et al.
Science (New York, N.Y.), 212(4498), 1038-1040 (1981-05-29)
The fouling marine mussel Mytilus edulis attaches itself to various substrates by spinning byssal threads, the adhesive discs of which are rich in the amino acid 3,4-dihydroxyphenylalanine (dopa). An acid-soluble protein was extracted and purified from the phenol gland located
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service