Skip to Content
Merck
All Photos(1)

Documents

763977

Sigma-Aldrich

Tri(ethylene glycol) diacrylate

97%, cross-linking reagent polymerization reactions, ≤1000 ppm MEHQ as inhibitor

Synonym(s):

Triethyleneglycol diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O6
CAS Number:
Molecular Weight:
258.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Tri(ethyleneglycol) diacrylate, contains ≤1000 ppm MEHQ as inhibitor, 97%

Assay

97%

form

liquid

contains

≤1000 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.464

density

1.110 g/mL at 25 °C

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

C=CC(=O)OCCOCCOCCOC(=O)C=C

InChI

1S/C12H18O6/c1-3-11(13)17-9-7-15-5-6-16-8-10-18-12(14)4-2/h3-4H,1-2,5-10H2

InChI key

INQDDHNZXOAFFD-UHFFFAOYSA-N

General description

Triethyleneglycol dimethacrylate is used as an intermediate product for polymer synthesis in the chemical industry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Van Miller et al.
Regulatory toxicology and pharmacology : RTP, 37(1), 54-65 (2003-03-29)
Specialty acrylates and methacrylates (SAM) comprise a large family of industrial monomers. In the late 1980s, the United States EPA and the industry SAM Panel collaborated to evaluate the potential effects, particularly carcinogenesis, of this family of chemicals. As part
Triethylene glycol diacrylate.
American Industrial Hygiene Association journal, 42(11), B47-B48 (1981-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service