Skip to Content
Merck
All Photos(1)

Documents

760137

Sigma-Aldrich

Benzo[1,2-b:4,5-b′]dithiophene-4,8-dione

97%

Synonym(s):

Thieno[2,3-f]benzothiophene-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4O2S2
CAS Number:
Molecular Weight:
220.27
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

260-265 °C

SMILES string

O=C1c2ccsc2C(=O)c3ccsc13

InChI

1S/C10H4O2S2/c11-7-5-1-3-13-9(5)8(12)6-2-4-14-10(6)7/h1-4H

InChI key

SIUXRPJYVQQBAF-UHFFFAOYSA-N

General description

Benzo[1,2-b:4,5-b′]dithiophene-4,8-dione (BDTD) is a conductive polymer with a symmetric and planar conjugated structure which has a hole mobility of 0.25 cm2V-1s-1.

Application

BDTD can be used as an acceptor layer in the fabrication of polymer based solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low bandgap polymers with benzo (1, 2-b: 4, 5-b?) dithiophene and bisthiophene-dioxopyrrolothiophene units for photovoltaic applications.
Zhang G, et al.
Polymer, 52(2), 415-421 (2011)
Yongye Liang et al.
Journal of the American Chemical Society, 131(22), 7792-7799 (2009-05-21)
This paper describes synthesis and photovoltaic studies of a series of new semiconducting polymers with alternating thieno[3,4-b]thiophene and benzodithiophene units. The physical properties of these polymers were finely tuned to optimize their photovoltaic effect. The substitution of alkoxy side chains
Optimization of side chains in alkylthiothiophene-substituted benzo (1, 2-b: 4, 5-b?) dithiophene-based photovoltaic polymers.
Zhang S, et al.
Polym. Chem., 6(14), 2752-2760 (2015)
Coffin; R. C.; MacNeill; C. M.; Peterson; E. D.; et al.
Journal of Nanotechnology, 572329-572329 (2011)
Synthesis and photovoltaic properties of low band gap polymers containing benzo (1, 2-b: 4, 5-c?) dithiophene-4, 8-dione.
Cao J, et al.
Macromolecules, 45(3), 1710-1714 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service