Skip to Content
Merck
All Photos(1)

Key Documents

755672

Sigma-Aldrich

Aluminum-scandium alloy

Al 98 wt. %, Sc 2 wt. %, 99.5% trace metals basis

Synonym(s):

Al-Sc, Aluminum scandium master alloy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.5% trace metals basis

form

ingot (20mmx25.4mmx25.4mm +/- 0.3mm)

composition

Al, 98 wt. %
Sc, 2 wt. %

mp

655 °C (lit.)

SMILES string

[AlH3].[Sc]

InChI

1S/Al.Sc.3H

InChI key

JJWKWQDCAMHJNV-UHFFFAOYSA-N

Related Categories

General description

Sc content 1.5-2.5 wt. %

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomus, D.; Qian, M.; Brice, C.A.; Muddle, B.C.
Scr. Mater., 63, 151-151 (2010)
Stock, J.-R.; Koehler, B.; Bomas, H.; Zoch, H.-W.
Materials & Design, 31, S76-S76 (2010)
Marquis, E.A.; Seidman, D.N.; Dunand, D.C.
Acta Materialia, 50, 4021-4021 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service