Skip to Content
Merck
All Photos(1)

Key Documents

753955

Sigma-Aldrich

ICBA

99% (HPLC)

Synonym(s):

1′,1′′,4′,4′′-Tetrahydro-di[1,4]methanonaphthaleno[1,2:2′,3′,56,60:2′′,3′′][5,6]fullerene-C60, C60 derivative, indene-C60 bisadduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H16
CAS Number:
Molecular Weight:
952.96
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

99% (HPLC)

form

solid

loss

0.5 wt. % TGA, 235 °C

mp

253-260 °C

Orbital energy

LUMO 3.67 eV (CV)

OPV Device Performance

ITO/PEDOT:PSS/P3HT: ICBA (1:1)/Al

  • Short-circuit current density (Jsc): 10.82 mA/cm2
  • Open-circuit voltage (Voc): 0.78 V
  • Fill Factor (FF): 0.56
  • Power Conversion Efficiency (PCE): 4.65 %

ITO/PEDOT:PSS/P3HT:ICBA(1.5:1)/LiF/Al
  • Short-circuit current density (Jsc): 10.53 mA/cm2
  • Open-circuit voltage (Voc): 0.83 V
  • Fill Factor (FF): 0.60
  • Power Conversion Efficiency (PCE): 5.26 %

Looking for similar products? Visit Product Comparison Guide

General description

ICBA is a C60 derivative that can be synthesized from C60 and indene. It is an acceptor material with shallow lowest occupied molecular orbital(LUMO) which can be used to enhance the performance of polymeric solar cells(PSCs).
The performance of indene-C60 bisadduct (ICBA) as organic solar cell (OSC) has been described in a study. Simple preparation and high solubility inorganic solvents renders ICBA as an efficient substitute to [6,6]-phenyl-C61 butyric acid methyl ester (PCBM).

Application

A ternary bulk heterojunction system (BHJ) may be fabricated with ICBA low band gap polymer, for IR sensitized organic solar cells. 1-3ICBA/thiazolo-thiazolo donor polymer blend may be used to enhance the charge generation in BHJ solar cells.
ICBA can be used with poly(3-hexylthiophene)(P3HT) as an active layer which can be used in the fabrication of organic solar cells(OSCs).ICBA in combination with thiazolo-thiazolo can be used as a donor. It can be blended with PCBM for photovoltaic based application.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of the structural configuration of ICBA/P3HT photovoltaic cells.
Nemnes GA, et al.
Applied Surface Science, 424(4), 264-268 (2017)
Temperature dependence of P3HT: ICBA polymer solar cells.
2017 International Conference on Numerical Simulation of Optoelectronic Devices, 133-134 (2017)
Photo-physics of PTB7, PCBM and ICBA based ternary solar cells.
Sharma R, et al.
Organic Electronics, 34(4), 111-117 (2016)
Charge Photogeneration for a Series of Thiazolo-Thiazole Donor Polymers Blended with the Fullerene Electron Acceptors PCBM and ICBA.
Shoaee S, et al.
Advances in Functional Materials, 23(26), 3286-3298 (2013)
Hyunbum Kang et al.
ACS applied materials & interfaces, 4(1), 110-116 (2011-12-14)
The ability to tune the lowest unoccupied molecular orbital (LUMO)/highest occupied molecular orbital (HOMO) levels of fullerene derivatives used as electron acceptors is crucial in controlling the optical/electrochemical properties of these materials and the open circuit voltage (V(oc)) of solar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service