Skip to Content
Merck
All Photos(2)

Documents

752525

Sigma-Aldrich

2-Hydroxyterephthalic acid

greener alternative

97%

Synonym(s):

2-Hydroxy-1,4-benzenedicarboxylic acid, H2BDC-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O5
CAS Number:
Molecular Weight:
182.13
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

312-317 °C

greener alternative category

SMILES string

OC(=O)c1ccc(C(O)=O)c(O)c1

InChI

1S/C8H6O5/c9-6-3-4(7(10)11)1-2-5(6)8(12)13/h1-3,9H,(H,10,11)(H,12,13)

InChI key

CDOWNLMZVKJRSC-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeru Kohtani et al.
Physical chemistry chemical physics : PCCP, 10(20), 2986-2992 (2008-05-14)
Superoxide anion radical (O(2)(-*)) and OH radical generations in suspensions of Ag metal-, Ag(2)O-, or AgO-loaded TiO(2) and BiVO(4) photocatalysts in alkaline conditions (pH 12.0) were examined by means of a luminol chemiluminescence (CL) technique and a spin-trapping fluorescence one
X Qu et al.
Photochemistry and photobiology, 71(3), 307-313 (2000-03-25)
The known photoreactions of melanin include production of melanin free radicals and oxygen consumption. While qualitative descriptions of the intermediates and products of these processes have been published, no quantitative procedures have been reported that allow the convenient measurement of
Yonggang Zhao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(18), 5101-5109 (2011-03-25)
Hydroxyl- and amino- functionalized [Zn(BDC)(TED)(0.5)]·2DMF·0.2H(2)O leads to two new structures, [Zn(BDC-OH)(TED)(0.5)]·1.5DMF·0.3H(2)O and [Zn(BDC-NH(2))(TED)(0.5)]·xDMF·yH(2)O (BDC=terephthalic acid, TED=triethylenediamine, BDC-OH=2-hydroxylterephthalic acid, BDC-NH(2)=2-aminoterephthalic acid). Single-crystal X-ray diffraction and powder X-ray diffraction studies confirmed that the structures of both functionalized compounds are very similar to
Sebastian Henke et al.
Journal of the American Chemical Society, 133(7), 2064-2067 (2011-02-01)
Covalent functionalization of 1,4-benzenedicarboxylate (bdc) with methoxyethoxy groups induces conformational freedom in this molecule. Applying these 2,5-disubstituted bdc derivatives in metal-organic framework synthesis together with 4,4'-bipyridine as coligand yields novel honeycomb-like structures. The cylindrical channels of these materials are populated
On the generation of free radical species from quantum dots.
Binil Itty Ipe et al.
Small (Weinheim an der Bergstrasse, Germany), 1(7), 706-709 (2006-12-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service