Skip to Content
Merck
All Photos(1)

Key Documents

745073

Sigma-Aldrich

N-[(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl]-1,8-diamino-3,6-dioxaoctane

for Copper-free Click Chemistry

Synonym(s):

(1α,8α,9β)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate, BCN-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28N2O4
CAS Number:
Molecular Weight:
324.42
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.22

form

liquid

composition

carbon content, 62.94%
hydrogen content, 8.70%
nitrogen content, 8.63%

reaction suitability

reaction type: click chemistry
reagent type: linker

solubility

water: 1.5 g/L

density

1.14  g/cm3 at 20  °C

functional group

amine

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12CCC#CCC[C@]1([H])[C@@H]2COC(NCCOCCOCCN)=O

InChI

1S/C17H28N2O4/c18-7-9-21-11-12-22-10-8-19-17(20)23-13-16-14-5-3-1-2-4-6-15(14)16/h14-16H,3-13,18H2,(H,19,20)/t14-,15+,16-

InChI key

YZGOWXGENSKDSE-MUJYYYPQSA-N

Application

Amine functionalized cyclooctyne derivative. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This strained cyclooctyne will react with azide functionalized compounds or biomolecules without the need for a copper catalyst to result in a stable triazole linkage.
N-[(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl]-1,8-diamino-3,6-dioxaoctane may be used to functionalize hyaluronic acid with bicyclo[6.1.0]nonyne (BCN) for the development of responsive HA-based physical hydrogels with tunable properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparison of strain-promoted alkyne-azide cycloaddition with established methods for conjugation of biomolecules to magnetic nanoparticles
Gruttner C, et al.
IEEE Transactions on Magnetics, 49(1), 172-176 (2013)
Jonathan Cortese et al.
Stroke, 52(2), 677-686 (2021-01-09)
Beyond aneurysmal occlusion, metallic flow diverters (FDs) can induce an adverse endovascular reaction due to the foreignness of metal devices, hampering FD endothelialization across the aneurysm neck, and arterial healing of intracranial aneurysms. Here, we evaluated the potential benefits of
Orientation of llama antibodies strongly increases sensitivity of biosensors
Trilling AK, et al.
Biosensors And Bioelectronics, 60, 130-136 (2014)
Jonas Christoffersson et al.
Biofabrication, 11(1), 015013-015013 (2018-12-14)
Liver cell culture models are attractive in both tissue engineering and for development of assays for drug toxicology research. To retain liver specific cell functions, the use of adequate cell types and culture conditions, such as a 3D orientation of
Christopher Aronsson et al.
Biofabrication, 12(3), 035031-035031 (2020-05-20)
Hydrogels are used in a wide range of biomedical applications, including three-dimensional (3D) cell culture, cell therapy and bioprinting. To enable processing using advanced additive fabrication techniques and to mimic the dynamic nature of the extracellular matrix (ECM), the properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service