Skip to Content
Merck
All Photos(1)

Key Documents

740683

Sigma-Aldrich

1,8-Dihydroxynaphthalene

95%

Synonym(s):

1,8-Naphthalenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
Beilstein:
2044947
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

137-143 °C

storage temp.

2-8°C

SMILES string

Oc1cccc2cccc(O)c12

InChI

1S/C10H8O2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6,11-12H

InChI key

OENHRRVNRZBNNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,8-Dihydroxynaphthalene (DHN) can be used as:
  • An intermediate in the preparation of benzo analogs of spiromamakone A.
  • A starting material to synthesize naphthopyran derivatives.
  • An intermediate in the total synthesis of palmarumycin CP17 analogs.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E S Jacobson et al.
Infection and immunity, 63(12), 4944-4945 (1995-12-01)
1,8-Dihydroxynaphthalene melanin in Wangiella dermatitidis and Alternaria alternata was titrated in vivo with the oxidants permanganate, hypochlorite, and H2O2. Melanized strains neutralized more oxidant and withstood higher concentrations of permanganate and hypochlorite than albino strains did. H2O2 killing required 1,000-fold
E Thines et al.
The Journal of antibiotics, 51(4), 387-393 (1998-06-19)
From submerged cultures of Scytalidium sp. 36-93, ten metabolites were isolated due to their effects on dihydroxynaphthalene (DHN) or DOPA melanin biosynthesis. Four of the compounds, scytalols A (1a), B (1b), C (2) and D (3), are new secondary metabolites
Pigment biosynthesis and virulence.
A A Brakhage et al.
Contributions to microbiology, 2, 205-215 (1999-10-16)
Bo Wang et al.
BMC genomics, 18(1), 729-729 (2017-09-17)
Ectophytic fungi occupy the waxy plant surface, an extreme environment characterized by prolonged desiccation, nutrient limitation, and exposure to solar radiation. The nature of mechanisms that facilitate adaptation to this environment remains unclear. In this study, we sequenced the complete
H F Tsai et al.
Journal of bacteriology, 180(12), 3031-3038 (1998-06-11)
Aspergillus fumigatus, an important opportunistic pathogen which commonly affects neutropenic patients, produces conidia with a bluish-green color. We identified a gene, alb1, which is required for conidial pigmentation. The alb1 gene encodes a putative polyketide synthase, and disruption of alb1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service